Methyl (R)-(-)-3-hydroxyisobutyrate ≥99.0%

Leverantör: TCI EUROPE

Synonyms: Methyl (R)-(-)-beta-hydroxyisobutyrate, (R)-Methyl 3-hydroxy-2-methylpropanoate

H0703-25G H0703-5G
TCIAH0703-25GEA 6300 SEK
TCIAH0703-25G TCIAH0703-5G
Methyl (R)-(-)-3-hydroxyisobutyrate ≥99.0%
Methyl (R)-(-)-3-hydroxyisobutyrate
Formel: C₅H₁₀O₃
MW: 118,1311 g/mol
Kokpunkt: 60°C
Flampunkt: 81°C
MDL Number: MFCD00063450
CAS nummer: 72657-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c8ced2b7bcff02f73acf8c04a540955a2bd2c6e&vid=2c8ced2b7bcff02f73acf8c04a540955a2bd2c6e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Optical purity (GC) min. 99.0 ee%
Specific rotation [a]20/D -22.0 to -23.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR