2-Hydrazinobenzothiazole ≥99.0%

Leverantör: TCI
Fara

Synonyms: 2-Hydrazinylbenzo[d]thiazole

H0177-25G H0177-5G
TCIAH0177-25GEA 1725 SEK
TCIAH0177-25G TCIAH0177-5G
2-Hydrazinobenzothiazole ≥99.0%
2-Hydrazinobenzothiazole
Formel: C₇H₇N₃S
MW: 165.21 g/mol
MDL Number: MFCD00041849
CAS nummer: 615-21-4
EINECS: 210-416-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dccaf8412922a5c3b9c786d178b196360ca1c53&vid=8dccaf8412922a5c3b9c786d178b196360ca1c53&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR