3-Nitro-1,8-naphthalic anhydride 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Nitrobenzo[de]isochromene-1,3-dione
278730050 278730250
ACRO278730050EA 900 SEK
ACRO278730050 ACRO278730250
3-Nitro-1,8-naphthalic anhydride 98%
3-Nitro-1,8-naphthalic anhydride
Formel: C₁₂H₅NO₅
MW: 243,1718 g/mol
Smältpunkt: 247…254 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006926
CAS nummer: 3027-38-1
Order Now
Specifikation testresultat
Appearance Light yellow to beige to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with TBAH ≥98.5 %