HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
H26082.03 H26082.06 H26082.14
H26082.03EA 885 SEK
H26082.03 H26082.06 H26082.14
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99%
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Purity: 99% CAS No.: 148893-10-1 Molecular Formula: C10H15F6N6OP Formula Weight: 380.23 Melting Point: ca 185° dec. MDL No.: MFCD00274639
Formel: C₁₀H₁₅F₆N₆OP
MW: 380.2 g/mol
Smältpunkt: ∼ 185 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00274639
CAS nummer: 148893-10-1
UN: 1325
ADR: 4.1 II