O-Methylisourea hemisulphate 94%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
O-Methylisouronium sulphate
233030250
233032500
ACRO233030250EA
1275
SEK
ACRO233030250
ACRO233032500
O-Methylisourea hemisulphate 94%
O-Methylisourea hemisulphate
Formel:
C₄H₁₄N₄O₆S MW: 246,24 g/mol Smältpunkt: 147…153 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00040594 CAS nummer: 52328-05-9 EINECS: 257-851-8 |
Specifikation testresultat
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with NaOH | ≥92 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...