1-(2-Tetrahydrofuroyl)piperazine ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone
T2617-25G
T2617-5G
TCIAT2617-25GEA
685
SEK
TCIAT2617-25G
TCIAT2617-5G
1-(2-Tetrahydrofuroyl)piperazine ≥97.0%
1-(2-Tetrahydrofuroyl)piperazine
Formel:
C₉H₁₆N₂O₂ MW: 184,24 g/mol Kokpunkt: 125°C |
MDL Number:
MFCD00190247 CAS nummer: 63074-07-7 |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...