1-(2-Tetrahydrofuroyl)piperazine ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone
L18704.14 L18704.06
L18704.14EA 2400 SEK
L18704.14 L18704.06
1-(2-Tetrahydrofuroyl)piperazine ≥97%
1-(2-Tetrahydrofuroyl)piperazine
Purity: 97% CAS No.: 63074-07-7 Molecular Formula: C9H16N2O2 Formula Weight: 184.24 Boiling Point: 120-125°/0.2mm Beilstein Reference: 880578
Formel: C₉H₁₆N₂O₂
MW: 184.24 g/mol
Kokpunkt: 120…125 °C (0.2 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00190247
CAS nummer: 63074-07-7