Estrone ≥98.0%

Leverantör: TCI
Fara

E0026-1G E0026-5G
TCIAE0026-1GEA 405 SEK
TCIAE0026-1G TCIAE0026-5G
Estrone ≥98.0%
Estrone
Formel: C₁₈H₂₂O₂
MDL Number: MFCD00003620
CAS nummer: 53-16-7
EINECS: 200-164-5
Merck Index: 14,03708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fe92e7a492c7016984f93f1436fd4e15879bcff&vid=7fe92e7a492c7016984f93f1436fd4e15879bcff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +161.0 to +168.0 deg (C=1, Dioxane)
Solubility in Dioxane very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR