Estriol ≥98.0%

Leverantör: TCI EUROPE
Fara

E0218-1G E0218-100MG
TCIAE0218-1GEA 1400 SEK
TCIAE0218-1G TCIAE0218-100MG
Estriol ≥98.0%
Estriol
Formel: C₁₈H₂₄O₃
MDL Number: MFCD00003691
CAS nummer: 50-27-1
EINECS: 200-022-2
Merck Index: 14,03707
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3a1971f19e65bdd3f64256d52073a8d565962a5&vid=d3a1971f19e65bdd3f64256d52073a8d565962a5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +55.0 to +60.0 deg (C=0.4, Dioxane)
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR