Zirkonium(IV)oxid ≥99.4% stabilised, pulver -100 Mesh
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Zirconium dioxide, Zirconium Oxide
Stabilization: 4% Stabilised with calcia (CaO)
40452.18 40452.A1
40452.18EA 1075 SEK
40452.18 40452.A1
Zirkonium(IV)oxid ≥99.4% stabilised, pulver -100 Mesh
Zirconium(IV) oxide
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.
Plasma Spray Grade. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, Alcohols, Halogens.
Formel: O₂Zr
MW: 123.22 g/mol
Kokpunkt: ∼ 5000 °C
Smältpunkt: ∼ 2700 °C
Densitet: 5.75 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180