Ethyl-3-phenylglycidate (mixture of cis and trans isomers) ≥90%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl 3-phenylglycidate
B22990.AD B22990.AK
B22990.ADEA 790 SEK
B22990.AD B22990.AK
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) ≥90%
Ethyl-3-phenylglycidate (mixture of cis and trans isomers)
Purity: 90+% CAS No.: 121-39-1 Molecular Formula: C11H12O3 Formula Weight: 192.21 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5180 Boiling Point: 95-97°/0.5mm MDL No.: MFCD00005123
Formel: C₁₁H₁₂O₃
MW: 192,21 g/mol
Kokpunkt: 95…97 °C (0,5 mmHg)
Densitet: 1,102
Flampunkt: >110 °C (230 °F)
MDL Number: MFCD00005123
CAS nummer: 121-39-1
EINECS: 204-467-3