Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), Co(TMHD)3
43993.02 43993.04 43993.09
43993.02EA 1500 SEK
43993.02 43993.04 43993.09
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)
Purity: 99% CAS No.: 14877-41-9 Molecular Formula: C33H57CoO6 Formula Weight: 608.72 Physical Form: Powder Appearance: Green Melting Point: 254-256° Boiling Point: subl. 120°/0.5mm MDL No.: MFCD00233617
Vapor deposition
Formel: C₃₃H₅₇CoO₆
MW: 608,735 g/mol
Kokpunkt: 120 °C (0,5 mmHg) (subl.)
Smältpunkt: 254…256 °C
Storage Temperature: Omgivande
MDL Number: MFCD00233617
CAS nummer: 14877-41-9