Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III) ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: La(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)
36465.03 36465.06
36465.03EA 2000 SEK
36465.03 36465.06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III) ≥98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III)
Purity: 98% CAS No.: 14319-13-2 Molecular Formula: C33H57LaO6 Formula Weight: 688.72 Physical Form: Powder Sensitivity: Hygroscopic Boiling Point: 370° dec. MDL No.: MFCD00010466
Formel: C₃₃H₅₇LaO₆
MW: 688,71 g/mol
Kokpunkt: 370 °C (dec.)
Smältpunkt: 240 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010466
CAS nummer: 14319-13-2