PLOT columns, CP-PoraPLOT Amines
Leverantör: VARIAN
CP7591 CP7594
VARICP7591EA 45000 SEK
VARICP7591 VARICP7594
PLOT columns, CP-PoraPLOT Amines
Kromatografikolonn
CP-PoraPLOT Amines is a unique column specially designed for the high retention analysis of very volatile amines.
  • Guaranteed performance for volatile amines providing ease-of-use
  • Very high efficiency at temperatures above ambient for lower cost per analysis
  • High sensitivity for amines and ammonia for accurate results
Temperature limits: -100 to 220/220 °C