CHROMABOND® C18 SPE Columns
Leverantör: MACHEREY-NAGEL
730130 730261 730400 730001 730003 730004 730005 730028 731801 731802 731803 738001.100M 730004.250 730003.250 730005.250
MANA730130EA 5825 SEK
MANA730130 MANA730261 MANA730400 MANA730001 MANA730003 MANA730004 MANA730005 MANA730028 MANA731801 MANA731802 MANA731803 MANA738001.100M MANA730004.250 MANA730003.250 MANA730005.250
CHROMABOND® C18 SPE Columns
Kromatografikolonn
CHROMABOND® C18 is an octadecyl modified silica phase for SPE.
  • Nonpolar
  • Hydrophobic and secondary interactions with polar groups of the analytes
  • Suitable for nonpolar and slight polar substances
Recommended for many nonpolar compounds with polar groups like pesticides, PCBs, pharmaceuticals and food ingredients.