Bottle top vacuum filtration systems
Leverantör: witeg Labortechnik
8.217.302.K
WITG8.217.302.KEA 12200 SEK
WITG8.217.302.K
Bottle top vacuum filtration systems
Filter Filter för Flaskor
The borosilicate glass vacuum filtration system with glass frit is complete with perforated PP screw thread cap GL 14, collecting flask 1000 ml, glass lower part with plastic hose connector, and seal elements PTFE.