Sedimentation cones, Imhoff type
Leverantör: witeg Labortechnik
2.936.000
WITG2.936.000EA 6425 SEK
WITG2.936.000
Sedimentation cones, Imhoff type
Trattar Sedimentationskärl
Made of borosilicate glass.
  • Graduated
  • With NS stopcock
  • Very good temperature and chemical resistance