Column head with valve stopcock acc. to Antlinger
Leverantör: WITEG LABORTECHNIK
1.610.029
WITG1.610.029EA 20950 SEK
WITG1.610.029
Column head with valve stopcock acc. to Antlinger
Destillationsutrustning Destillationsuppsats
Borosilicate glass3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Columns heads acc. to Antlinger, with PTFE-fine-dosing valve, cone to cloumn NS 29/32.