Distilling heads, Claisen type
Leverantör: witeg Labortechnik
1.020.004
WITG1.020.004EA 1675 SEK
WITG1.020.004
Distilling heads, Claisen type
Destillationsutrustning Destillationsuppsats
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Acc. to DIN 12594 and in addition to standard, with two parallel NS sockets and two NS cones (descending angle: 75°), for distillation thermometers of 55 mm built-in length.