WHEATON® I-Loc™ Polypropylene, Screw Thread, Closure, with, Stopper
Leverantör: DWK Life Sciences
W010342J
WHEAW010342JEA 0 SEK
WHEAW010342J
WHEATON® I-Loc™ Polypropylene, Screw Thread, Closure, with, Stopper
Lock Lock, flaskor
For use with screw thread bottles and vials.
  • Siliconized grey bromobutyl stopper, 50 Durometer
  • Autoclavable
Polypropylene open top screw closure for piercing applications.