WHEATON® MicroLiter® 76 Vials with Limited Volume Inserts, Clear, Silanized
Leverantör: DWK Life Sciences
76-1000-101-S
WHEA76-1000-101-SEA 4150 SEK
WHEA76-1000-101-S
WHEATON® MicroLiter® 76 Vials with Limited Volume Inserts, Clear, Silanized
Vialer
76-Series Vials™ have widest opening for ease of sample addition, sample removal and universal autosampler compatibility. Certified Class 10,000 clean room packed enhances data integrity.
Certifikat: Certified Class 10,000.