Stoppers for BOD bottles, WHEATON®
Leverantör: DWK Life Sciences
227670 227672
WHEA227670EA 1725 SEK
WHEA227670 WHEA227672
Stoppers for BOD bottles, WHEATON®
Proppar Glas
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For 60 ml and 300 ml BOD bottles.
Order Now
SPECIFIKATIONER
Environmentally Preferable