Gas-safety adapter 360 for Flame 100
Leverantör: WARTEWIG
2.000.701
WART2.000.701EA 1650 SEK
WART2.000.701
Gas-safety adapter 360 for Flame 100
Värmeapparater Brännare, gas
Gas-safety adapter 360 for Flame 100.
  • With integrated pressure regulator
  • Continuous cartridge operation 65 minutes
Ideal for cartridges CV 360, 52 g from Camping gas.