Colorimeter, Libra S2
Leverantör: Biochrom
80-5000-02
WAPR80-5000-02EA 0 SEK
WAPR80-5000-02
Colorimeter, Libra S2
Kolorimetrar Kolorimetrar, Multiparameter
The Libra S2 is a small, robust, easy to use instrument that has been designed with both the student and field user in mind. It is ideal for teaching principles of science and analysis in schools and colleges.
  • Rechargeable NiMH batteries
  • Extremely versatile and ideal for QC and educational applications
  • Instrument surface is designed for ease of cleaning and decontamination
The instrument measures in absorbance and % transmission mode as well as in simple kinetics, enabling changes in absorbance over time and reaction rates to be determined. TheMore Product Information