Sani-Tech® STHT®-R Hoses
Leverantör: Saint Gobain Life Sciences
STHT-R-0250 STHT-R-0750 STHT-R-0500 STHT-R-0125 STHT-R-0125F STHT-R-0125-DB STHT-R-0125F-D STHT-R-0375 STHT-R-0250F STHT-R-0250-DB STHT-R-0250F-D STHT-R-0375F STHT-R-0375-DB STHT-R-0375F-D STHT-R-0500F STHT-R-0500F-D STHT-R-0625 STHT-R-0625F STHT-R-0625-DB STHT-R-0625F-D STHT-R-0750F STHT-R-0750-DB STHT-R-0750F-D STHT-R-0875 STHT-R-0875F STHT-R-0875-DB STHT-R-087F5-D STHT-R-1000 STHT-R-1000F STHT-R-1000-DB STHT-R-1000F-D
VERNSTHT-R-0250EA 16125 SEK
VERNSTHT-R-0250 VERNSTHT-R-0750 VERNSTHT-R-0500 VERNSTHT-R-0125 VERNSTHT-R-0125F VERNSTHT-R-0125-DB VERNSTHT-R-0125F-D VERNSTHT-R-0375 VERNSTHT-R-0250F VERNSTHT-R-0250-DB VERNSTHT-R-0250F-D VERNSTHT-R-0375F VERNSTHT-R-0375-DB VERNSTHT-R-0375F-D VERNSTHT-R-0500F VERNSTHT-R-0500F-D VERNSTHT-R-0625 VERNSTHT-R-0625F VERNSTHT-R-0625-DB VERNSTHT-R-0625F-D VERNSTHT-R-0750F VERNSTHT-R-0750-DB VERNSTHT-R-0750F-D VERNSTHT-R-0875 VERNSTHT-R-0875F VERNSTHT-R-0875-DB VERNSTHT-R-087F5-D VERNSTHT-R-1000 VERNSTHT-R-1000F VERNSTHT-R-1000-DB VERNSTHT-R-1000F-D
Sani-Tech® STHT®-R Hoses
Slangar Överföringsslang
Sani-Tech® STHT®-R braid-reinforced, platinum-cured silicone hose is an ultra-flexible, high-purity hose that was developed for higher-pressure applications. It is manufactured with Sani-Tech® 65 custom-brand silicone resin. Sani-Tech STHT-R hose resists temperature extremes, ozone, radiation, moisture, compression sets, weathering, and chemical attack. It imparts no taste or odours to fluids transported.
  • Biopharmaceutical-grade silicone hose
  • High pressure ratings
  • Ultra-flexible
  • Improved bend radius over wire-reinforced hose
  • Autoclavable and sterilisable
More Product Information