SIPS 10/20 pump tubing
Leverantör: VARIAN
9910076100
VARI9910076100EA 1850 SEK
VARI9910076100
SIPS 10/20 pump tubing
Slangar Peristaltisk pumpslang
SIPS 10/20 pump tubing for atomic absorption spectroscopy.
  • Selected to provide extended lifetime and accurate dilution when using the SIPS
The Sample Introduction Pump System (SIPS) is an innovative sample introduction and dilution system for flame AA that eliminates the task of manually preparing multiple calibration standards, providing fast, accurate on-line dilution of over range samples.