Laser Protection Goggles
Leverantör: UNIVET
562S.00.01.312
UNVT562S.00.01.312EA 4500 SEK
UNVT562S.00.01.312
Laser Protection Goggles
Skyddsglasögon
  • Filter: Full protection
  • VLT: 40%
  • Alignement laser wavelength T%(λ) >10%: 410 to 695 nm