T-Classic Plus Gauged Autoclave
Leverantör: TUTTNAUER
210001XP 210004XP 210048XP 210052XP
TUTT210001XPEA 16875 SEK
TUTT210001XP TUTT210004XP TUTT210048XP TUTT210052XP
T-Classic Plus Gauged Autoclave
Sterilisatorer Autoklaver (ångsterilisering)
The T-Classic plus gauged autoclave is ideal for effective sterilization at a higher temperature. Complete with gauges to show pressure and temperature, this autoclave is designed for laboratory use and with its simple one touch operation, it is guaranteed to meet your sterilization needs.
  • Lightweight and portable
  • Intutive and simple
  • Compact and safe
  • 9 and 12 litre capacity
  • Sterilizes at 126 °C
  • Gauges for temperature and pressure
The T-Classic Lab line includes portable steam autoclaves designed as a cost-effective sterilization solution. FullyMore Product Information