(-)-Verbenone ≥95.0%

Leverantör: TCI

Synonyms: (-)-2-Pinen-4-one, (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one, (-)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one, (1S)-(-)-Verbenone

V0072-25G
TCIAV0072-25GEA 895 SEK
TCIAV0072-25G
(-)-Verbenone ≥95.0%
(-)-Verbenone
Formel: C₁₀H₁₄O
MW: 150.22 g/mol
Flampunkt: 109°C
MDL Number: MFCD00001343
CAS nummer: 1196-01-6
EINECS: 214-807-2

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR