1-(2-Tetrahydrofurfuryl)-2-thiourea 98,0 HPLC, N

Leverantör: TCI
Fara

Synonyms: [(Tetrahydrofuran-2-yl)methyl]thiourea

T3413-200MG
TCIAT3413-200MGEA 2125 SEK
TCIAT3413-200MG
1-(2-Tetrahydrofurfuryl)-2-thiourea 98,0 HPLC, N
1-(2-Tetrahydrofurfuryl)-2-thiourea
Formel: C₆H₁₂N₂OS
MW: 160.24 g/mol
MDL Number: MFCD00060498
CAS nummer: 66892-25-9
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc412be90796113352725dc40753be7cc4b60178&vid=8a564e2572f65d8a0d9cc3d08b4b66a0bc553d95&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR