Tetrabutylgermane 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: Tetra-n-butylgermane, Tetra-n-butylgermanium

T1156-1G
TCIAT1156-1GEA 575 SEK
TCIAT1156-1G
Tetrabutylgermane 98,0 GC_ANALYSMETOD
Tetrabutylgermane
Formel: C₁₆H₃₆Ge
MW: 301.07 g/mol
Kokpunkt: 161°C
Storage Temperature: Omgivande
MDL Number: MFCD00015224
CAS nummer: 1067-42-1
EINECS: 213-929-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02028b0d0cd12467677a21bc659c8a7c7d337c69&vid=02028b0d0cd12467677a21bc659c8a7c7d337c69&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR