1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥98.0%

Leverantör: TCI
Varning

Synonyms: 3,4-Dihydronaphthalen-1(2H)-one

T0134-25G T0134-500G
TCIAT0134-25GEA 230 SEK
TCIAT0134-25G TCIAT0134-500G
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥98.0%
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone)
Formel: C₁₀H₁₀O
MW: 146.19 g/mol
Kokpunkt: 257°C
Flampunkt: 129°C
MDL Number: MFCD00001688
CAS nummer: 529-34-0
EINECS: 208-460-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40366ca64373bea589fe3011733d72c94455172a&vid=40366ca64373bea589fe3011733d72c94455172a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR