N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥96.0% (by HPLC, titration analysis)

Leverantör: TCI

Synonyms: 2,5-Dioxopyrrolidin-1-yl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate

S0866-1G S0866-200MG
TCIAS0866-1GEA 5925 SEK
TCIAS0866-1G TCIAS0866-200MG
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥96.0% (by HPLC, titration analysis)
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate
Formel: C₁₄H₉NO₇
MW: 303.23 g/mol
Storage Temperature: Frys
MDL Number: MFCD00037568
CAS nummer: 134471-24-2

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR