Standard mixture of n-Paraffins (consists of C17, C18, C19 and C20), standard material for GC
Leverantör: TCI
S0456-5G
TCIAS0456-5GEA 1150 SEK
TCIAS0456-5G
Standard mixture of n-Paraffins (consists of C17, C18, C19 and C20), standard material for GC
Standarder Petroleum Industry Standards