Quercetin hydrate 96,0 HPLC, UV

Leverantör: TCI
Fara

Synonyms: 3,3',4',5,7-Pentahydroxyflavone hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one xhydrate

P0042-25G
TCIAP0042-25GEA 695 SEK
TCIAP0042-25G
Quercetin hydrate 96,0 HPLC, UV
Quercetin hydrate
Formel: C₁₅H₁₀O₇·xH₂O
MW: 302.24 g/mol (anhy)
MDL Number: MFCD00006828
CAS nummer: 849061-97-8
EINECS: 204-187-1
UN: 2811
ADR: 6.1 III
Merck Index: 14,08034

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (UV-vis method) min. 96.0 % (calcd.on anh.substance)
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR