Naftalen 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

N0885-25G N0885-500G
TCIAN0885-25GEA 215 SEK
TCIAN0885-25G TCIAN0885-500G
Naftalen 98,0 GC_ANALYSMETOD
Naftalen
Formel: C₁₀H₈
MW: 128.17 g/mol
Kokpunkt: 218°C
Flampunkt: 79
Storage Temperature: Omgivande
MDL Number: MFCD00001742
CAS nummer: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1 III
Merck Index: 14,06370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4bd66d590350fdf511b0ab490eaee645121dc27&vid=e4bd66d590350fdf511b0ab490eaee645121dc27&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR