4-Methyl-3-nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Fara

Synonyms: 4-Methyl-3-nitrobenzene-1-sulphonyl chloride

N0704-25G N0704-5G
TCIAN0704-25GEA 9000 SEK
TCIAN0704-25G TCIAN0704-5G
4-Methyl-3-nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
4-Methyl-3-nitrobenzenesulphonyl chloride
Formel: C₇H₆ClNO₄S
MW: 235.65
Kokpunkt: 154°C
MDL Number: MFCD00129811
CAS nummer: 616-83-1
UN: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8440ac231e08bdb87d7446dd6d97c93567dcb7f0&vid=8440ac231e08bdb87d7446dd6d97c93567dcb7f0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 32.0 to 36.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR