2-Nitrobenzoic acid 95,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

N0155-25G N0155-500G
TCIAN0155-25GEA 485 SEK
TCIAN0155-25G TCIAN0155-500G
2-Nitrobenzoic acid 95,0 GC_ANALYSMETOD
2-Nitrobenzoic acid
Formel: C₇H₅NO₄
MW: 167.12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007137
CAS nummer: 552-16-9
EINECS: 209-004-9
Merck Index: 14,06588
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=727600fdb223918633fd54772eed8e5c1774ea98&vid=8217369ebdec013c6b9ce7562ff1a8688ec91bbb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR