1-Boc-isonipecotic acid methyl ester 98,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate

M2227-1G M2227-5G
TCIAM2227-1GEA 310 SEK
TCIAM2227-1G TCIAM2227-5G
1-Boc-isonipecotic acid methyl ester 98,0 GC_ANALYSMETOD
1-Boc-isonipecotic acid methyl ester
Formel: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
MDL Number: MFCD02183584
CAS nummer: 124443-68-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 33.0 to 37.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR