N-Methyl-N-(2-pyridyl)formamide 98,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: 2-(N-Formyl-N-methylamino)pyridine, Meyer's Reagent

M1187-1G M1187-5G
TCIAM1187-1GEA 550 SEK
TCIAM1187-1G TCIAM1187-5G
N-Methyl-N-(2-pyridyl)formamide 98,0 GC_ANALYSMETOD
N-Methyl-N-(2-pyridyl)formamide
Formel: C₇H₈N₂O
MW: 136.15 g/mol
Flampunkt: 109°C
MDL Number: MFCD00006260
CAS nummer: 67242-59-5
EINECS: 266-616-9

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR