(R)-(-)-Mandelic acid 99,0 GC, T

Leverantör: TCI
Varning

Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid

M0662-25G M0662-500G M0662-100G
TCIAM0662-25GEA 455 SEK
TCIAM0662-25G TCIAM0662-500G TCIAM0662-100G
(R)-(-)-Mandelic acid 99,0 GC, T
(R)-(-)-Mandelic acid
Formel: C₈H₈O₃
MW: 152.15 g/mol
Kokpunkt: 322
MDL Number: MFCD00064251
CAS nummer: 611-71-2
EINECS: 210-276-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=644a09de7bbeb4da749535736d2f891f12d2e57a&vid=644a09de7bbeb4da749535736d2f891f12d2e57a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 99.0 %
Melting point 132.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR