Hexamethoxydisiloxane 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: Hexamethyl Diorthosilicate

H1716-1G
TCIAH1716-1GEA 1475 SEK
TCIAH1716-1G
Hexamethoxydisiloxane 98,0 GC_ANALYSMETOD
Hexamethoxydisiloxane
Formel: C₆H₁₈O₇Si₂
MW: 258.37 g/mol
Kokpunkt: 73°C
MDL Number: MFCD02912820
CAS nummer: 4371-91-9
EINECS: 224-467-7
UN: 1993
ADR: 3 III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR