trans-2-Hexenal 97,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

Synonyms: (E)-2-Hexenal, trans-2-Hexenal

H0345-25ML H0345-500ML H0345-100ML
TCIAH0345-25MLEA 230 SEK
TCIAH0345-25ML TCIAH0345-500ML TCIAH0345-100ML
trans-2-Hexenal 97,0 GC_ANALYSMETOD
trans-2-Hexenal
Formel: C₆H₁₀O
MW: 98.14 g/mol
Kokpunkt: 146°C
Flampunkt: 43°C
Storage Temperature: Kylskåp
MDL Number: MFCD00007008
CAS nummer: 6728-26-3
EINECS: 229-778-1
UN: 1988
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71678219c2ca5a9177b78b5eac4d85ab62200edb&vid=71bf61ca4ec808310e7876b2e646d2bd4942e102&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR