4-[[(4-Fluorophenyl)imino]methyl]phenol 98,0 GC, T

Leverantör: TCI

Synonyms: ALPHA-(4-FLUOROPHENYLIMINO)-P-CRESOL, 4-[[(4-Fluorophenyl)imino]methyl]phenol

F0946-5G F0946-25G
TCIAF0946-5GEA 725 SEK
TCIAF0946-5G TCIAF0946-25G
4-[[(4-Fluorophenyl)imino]methyl]phenol 98,0 GC, T
4-[[(4-Fluorophenyl)imino]methyl]phenol
Formel: C₁₃H₁₀FNO
MW: 215.23 g/mol
MDL Number: MFCD00029739
CAS nummer: 3382-63-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR