1-Ethynyl-3,5-dimethoxybenzene 98,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: 3,5-Dimethoxyphenylacetylene

E1175-200MG E1175-1G
TCIAE1175-200MGEA 485 SEK
TCIAE1175-200MG TCIAE1175-1G
1-Ethynyl-3,5-dimethoxybenzene 98,0 GC_ANALYSMETOD
1-Ethynyl-3,5-dimethoxybenzene
Formel: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD03839985
CAS nummer: 171290-52-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR