Ethyl 3-Oxo-3-(2-thienyl)propionate 98,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: Ethyl 3-oxo-3-(thiophen-2-yl)propanoate, 3-Oxo-3-(2-thienyl)propionic acid Ethyl ester

E1073-200MG E1073-1G
TCIAE1073-200MGEA 325 SEK
TCIAE1073-200MG TCIAE1073-1G
Ethyl 3-Oxo-3-(2-thienyl)propionate 98,0 GC_ANALYSMETOD
Ethyl 3-Oxo-3-(2-thienyl)propionate
Formel: C₉H₁₀O₃S
MW: 198.24 g/mol
Kokpunkt: 130°C
MDL Number: MFCD00542649
CAS nummer: 13669-10-8

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR