N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine 98,0 HPLC, T

Leverantör: TCI

Synonyms: N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, NEPA, N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester, (S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoic acid, N-(1S)-Ethoxycarbonyl-3-phenylpropyl-(S)-alanine

E0530-25G E0530-5G
TCIAE0530-25GEA 1025 SEK
TCIAE0530-25G TCIAE0530-5G
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine 98,0 HPLC, T
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
Formel: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00209976
CAS nummer: 82717-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cea096c4fa2d835ced4ed2f7001f60abdabc37d4&vid=cea096c4fa2d835ced4ed2f7001f60abdabc37d4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 148.0 to 154.0 deg-C
Specific rotation [a]20/D +28.0 to +31.0 deg (C=1, Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR