2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥99.0%

Leverantör: TCI
Varning

Synonyms: 6-(2-(2-Undecyl-1H-imidazol-1-yl)ethyl)-1,3,5-triazine-2,4-diamine

D3239-25G
TCIAD3239-25GEA 390 SEK
TCIAD3239-25G
2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine ≥99.0%
2,4-Diamino-6-[2-(2-undecyl-1-imidazolyl)ethyl]-1,3,5-triazine
Formel: C₁₉H₃₃N₇
MW: 359.52 g/mol
MDL Number: MFCD00047352
CAS nummer: 50729-75-4
EINECS: 256-742-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 191.0 to 195.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR