(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol 98,0 HPLC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: (R)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diol

D2729-1G
TCIAD2729-1GEA 1950 SEK
TCIAD2729-1G
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol 98,0 HPLC_ANALYSMETOD
(R)-(-)-6,6'-Dibromo-1,1'-bi-2-naphthol
Formel: C₂₀H₁₂Br₂O₂
MW: 444.12 g/mol
MDL Number: MFCD00798290
CAS nummer: 65283-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2408a6c8ae222bc5789e7c25e9de6ad9e3a1ff0&vid=a2408a6c8ae222bc5789e7c25e9de6ad9e3a1ff0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D -44.0 to -50.0 deg (C=2,THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR