Etyldimetylamin 98,0 GC, T

Leverantör: TCI
Fara

Synonyms: DMEA

D1538-25ML D1538-500ML
TCIAD1538-25MLEA 215 SEK
TCIAD1538-25ML TCIAD1538-500ML
Etyldimetylamin 98,0 GC, T
Etyldimetylamin
Formel: C₄H₁₁N
MW: 73.14 g/mol
Kokpunkt: 37°C
Smältpunkt: -140
Flampunkt: –25°C
MDL Number: MFCD00009039
CAS nummer: 598-56-1
EINECS: 209-940-8
UN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f86f0977fc835435d77a0e5d50702dd07c91929b&vid=f86f0977fc835435d77a0e5d50702dd07c91929b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR